Caps for sample tubes, tamper evident
Lip seal screw cap, not tamper evident, 16,6 mm, White
SIMPT552W
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
- For tubes:16,6 mm
- Colour:White
- Description:Lip seal screw cap, not tamper evident
- Seq:05