Order Entry
Denmark
ContactUsLinkComponent
Stirrer head for PTFE-jacketed stirrer shaft dia.(d) 10 mm. Height (H) 145 mm. For ground joint NS 45/40. Insertion length of shaft (L2) max. 95 mm. Drive shaft O.D. (D) 50 mm. 1 * 1 items
Catalog # BOHLC530-15
Supplier:  Bohlender
undefined
Stirrer head for PTFE-jacketed stirrer shaft dia.(d) 10 mm. Height (H) 145 mm. For ground joint NS 45/40. Insertion length of shaft (L2) max. 95 mm. Drive shaft O.D. (D) 50 mm. 1 * 1 items
Catalog # BOHLC530-15
Supplier:  Bohlender
Frequently Bought Together