TemperatureProbes Lemo Compact EX (PT 100), One measuring sensor PT 100 in a stainless steel tube (1.4571) encapsulated with static dissipative PTFE-EX. Temperatureprobe Ø 8 mm, tip Ø 6 mm, collar ring Ø 12 mm. Electrostatic charges can be discharg 1 * 1 items
Catalog # BOHLP1734-15
Leverandør: Bohlender
Frequently Bought Together