Dispenser ceramus 0.4-2.0 ml conformity certifiedPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121° C without dismantling 1 * 1 items
Catalog # HIRS9322001
Supplier: HIRSCHMANN
Frequently Bought Together