Order Entry
Denmark
Orders LinkContactUsLinkComponent
DIN adapters for electrodes
DIN adapters for electrodes
DIN 19262 adapter T kit including temperature for LE438 / LE410
Catalog # METT30499900
Supplier:   Mettler - Toledo
DIN adapters for electrodes
Catalog # METT30499900
Supplier:  Mettler - Toledo
DIN adapters for electrodes
DIN 19262 adapter T kit including temperature for LE438 / LE410
Price
414,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These adapters will allow the LE electrodes with BNC to connect to meters with DIN.
Specifications
  • Description:
    DIN 19262 adapter T kit including temperature for LE438 / LE410