Order Entry
Denmark
Orders LinkContactUsLinkComponent
DIN adapters for electrodes
DIN adapters for electrodes
DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427
Catalog # METT30403890
Supplier:   Mettler - Toledo
DIN adapters for electrodes
Catalog # METT30403890
Supplier:  Mettler - Toledo
DIN adapters for electrodes
DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427
Price
283,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These adapters will allow the LE electrodes with BNC to connect to meters with DIN.
Specifications
  • Description:
    DIN 19262 adapter for LE407 / LE409 / LE420 / LE422 / LE427