A C1orf31 (Cytochrome c oxidase assembly factor 6 homolog) ELISA with a senstivity of 0.188ng/ml and a range of 0.313-20ng/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8182
Supplier: G-Biosciences
Frequently Bought Together