Order Entry
Denmark
ContactUsLinkComponent
Trichloroacetic acid (TCA) is used as a reagent for the precipitation of proteins and nucleic acids. Has been used to determine prot ein concentration by quantitative precipitation with the Lowry method. 1 * 500 g
Catalog # GENORC-813
Supplier:  G-Biosciences
Trichloroacetic acid (TCA) is used as a reagent for the precipitation of proteins and nucleic acids. Has been used to determine prot ein concentration by quantitative precipitation with the Lowry method. 1 * 500 g
Catalog # GENORC-813
Supplier:  G-Biosciences
Frequently Bought Together