Order Entry
Denmark
ContactUsLinkComponent
An ELISA kit for the detection of IgA (Monkey). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375ng/ml. 1 * 1 KIT
Catalog # GENOIT4803
Supplier:  G-Biosciences
An ELISA kit for the detection of IgA (Monkey). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375ng/ml. 1 * 1 KIT
Catalog # GENOIT4803
Supplier:  G-Biosciences
Frequently Bought Together