Order Entry
Denmark
ContactUsLinkComponent
An ELISA kit for the detection of LDHB (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 1.875ng/ml. 1 * 1 KIT
Catalog # GENOIT4099
Supplier:  G-Biosciences
An ELISA kit for the detection of LDHB (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 1.875ng/ml. 1 * 1 KIT
Catalog # GENOIT4099
Supplier:  G-Biosciences
Frequently Bought Together