Order Entry
Denmark
ContactUsLinkComponent
An ELISA kit for the detection of MCP-1 (Canine). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375pg/ml. 1 * 1 KIT
Catalog # GENOIT1242
Supplier:  G-Biosciences
An ELISA kit for the detection of MCP-1 (Canine). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375pg/ml. 1 * 1 KIT
Catalog # GENOIT1242
Supplier:  G-Biosciences
Frequently Bought Together