Order Entry
Denmark
ContactUsLinkComponent
GreenGel cup 3% TBE. Re-meltable agarose gel with fluorescent DNA stain in TBE buffer for 3% gels. Includes also 100ml TBE concentra te. 1 * 10 items
Catalog # EMBIM3123TBE
Supplier:  EMBITEC
GreenGel cup 3% TBE. Re-meltable agarose gel with fluorescent DNA stain in TBE buffer for 3% gels. Includes also 100ml TBE concentra te. 1 * 10 items
Catalog # EMBIM3123TBE
Supplier:  EMBITEC
Frequently Bought Together