Order Entry
Denmark
ContactUsLinkComponent
Urea Pretreat (compensates for the interference of free Chlorine up to 2 mg/l) 1 * 100 Tablet
Catalog # LOVI516110BT
Supplier:  Lovibond
Urea Pretreat (compensates for the interference of free Chlorine up to 2 mg/l) 1 * 100 Tablet
Catalog # LOVI516110BT
Supplier:  Lovibond
Frequently Bought Together