Order Entry
Denmark
ContactUsLinkComponent
A1BG (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human alpha 1B-glycoprotein in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5144
Supplier:  Abnova
A1BG (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human alpha 1B-glycoprotein in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5144
Supplier:  Abnova
Frequently Bought Together