Order Entry
Denmark
ContactUsLinkComponent
Kit containing MACS Equipment and Reagent for magnetic cell separation directly from whole blood using Whole Blood Columns 1 * 2 mL
Catalog # MILT130-098-240
Supplier:  MILTENYI BIOTEC
Kit containing MACS Equipment and Reagent for magnetic cell separation directly from whole blood using Whole Blood Columns 1 * 2 mL
Catalog # MILT130-098-240
Supplier:  MILTENYI BIOTEC
Frequently Bought Together