Order Entry
Denmark
ContactUsLinkComponent
Anti-Red Blood Cell (RBC) MicroBeads, rat were developed for the positive selection or depletion of rat erythrocytes e.g. from rat l ung, heart, fat, muscle or spleen as an alternative method to osmotic lysis. 1 * 1 items
  MILT130-109-681
Anti-Red Blood Cell (RBC) MicroBeads, rat were developed for the positive selection or depletion of rat erythrocytes e.g. from rat l ung, heart, fat, muscle or spleen as an alternative method to osmotic lysis. 1 * 1 items
  MILT130-109-681
Frequently Bought Together