Order Entry
Denmark
Orders LinkContactUsLinkComponent
Apigenin 97+%

Apigenin 97+%

Bottle, 25 g
Catalog # APOSBIK9018-25G
Irriterende
Supplier:   Apollo Scientific
Apigenin 97+%
Catalog # APOSBIK9018-25G
Supplier:  Apollo Scientific
CAS Number:  520-36-5
Apigenin 97+%

Bottle, 25 g

CAS Number:  520-36-5
Price
1.540,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
  • CAS nummer:520-36-5
  • Formel:C₁₅H₁₀O₅
  • MDL nummer:MFCD00006831
  • Molecular Weight:270.24 g/mol
  • Smeltende Pt:300 °C
  • Opbevaringstemperatur:Refrigerator
  • EINECS:208-292-3

Documents

Specifikationer

Specifikationer

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.