Order Entry
Denmark
Orders LinkContactUsLinkComponent
Aluminon

Aluminon

Bottle, 100 g
Catalog # APOSOR2429-100G
Irritant
Supplier:   Apollo Scientific
Aluminon
Catalog # APOSOR2429-100G
Supplier:  Apollo Scientific
CAS Number:  569-58-4
Aluminon

Bottle, 100 g

CAS Number:  569-58-4
Price
165,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
  • CAS Number:569-58-4
  • Formula:C₂₂H₂₃N₃O₉
  • MDL Number:MFCD00040925
  • Molecular Weight:473.44 g/mol
  • Melting Pt:220…225 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:209-319-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications