Order Entry
Denmark
Orders LinkContactUsLinkComponent
Storage Cabinet

Storage Cabinet

Storage cabinets, 2 doors, Stainless steel 304, 70 kg
Catalog # PALBJE0950C
Supplier:   Palbam
Storage Cabinet
Catalog # PALBJE0950C
Supplier:  Palbam
Storage Cabinet

Storage cabinets, 2 doors, Stainless steel 304, 70 kg

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Removable shelves and sloping roof design as standard. Available on either casters or leveler legs.

  • 3/4 height cabinets
  • stainless steel 304 with electro-polished finish

L×W×H: 900×500×1500 mm

Specifications
  • Description:
    Storage cabinets
  • Material:
    Stainless steel 304
  • Seq:
    01
  • Doors:
    2 doors
  • Weight:
    70 kg