Order Entry
Denmark
ContactUsLinkComponent
Threaded Couplings T, PTFE, PPS. For connecting two GL threads 18 (of the same size). Length from sealing lip to sealing lip 39 mm 1 * 1 items
Katalog # BOHLH908-18
Supplier:  Bohlender
undefined
Threaded Couplings T, PTFE, PPS. For connecting two GL threads 18 (of the same size). Length from sealing lip to sealing lip 39 mm 1 * 1 items
Katalog # BOHLH908-18
Supplier:  Bohlender
Frequently Bought Together