Order Entry
Denmark
ContactUsLinkComponent
Each impact-resistant 50 mL tube contains 1 mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae. 1 * 500 mL
Catalog # 116952010.
Supplier:  MP Biomedicals
Each impact-resistant 50 mL tube contains 1 mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae. 1 * 500 mL
Catalog # 116952010.
Supplier:  MP Biomedicals
Frequently Bought Together