Order Entry
Denmark
ContactUsLinkComponent
VISOCOLOR ECO Detergents cationic test kit - refill pack - measuring range: 1-20 mg/L CTAB sufficient for 50 determinations __UN 3316 Chemical kit 9 II 0.100 L ADR/GGVSEB M11 (E) ADR 3.3.1/251: LQ PAX 960 10 Kg CAO 960 10 Kg 1 * 1 items
Catalog # MANA931251
Supplier:  MACHEREY-NAGEL
undefined
VISOCOLOR ECO Detergents cationic test kit - refill pack - measuring range: 1-20 mg/L CTAB sufficient for 50 determinations __UN 3316 Chemical kit 9 II 0.100 L ADR/GGVSEB M11 (E) ADR 3.3.1/251: LQ PAX 960 10 Kg CAO 960 10 Kg 1 * 1 items
Catalog # MANA931251
Supplier:  MACHEREY-NAGEL