Order Entry
Denmark
Orders LinkContactUsLinkComponent
Screw caps
Screw caps
65 mm, 1000/1500/2000 ml
Catalog # 215-4246
Supplier:   Kautex
Screw caps
Catalog # 215-4246
Supplier:  Kautex
Screw caps
Screw caps
65 mm, 1000/1500/2000 ml
Price
1.450,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
LDPE, natural.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
Specifications
  • Thread:
    65 mm
  • Seq:
    06
  • For bottles:
    1000/1500/2000 ml