Screw caps
32 mm, 50/100 ml
Catalog # 215-4243
Supplier: Kautex
Jump to:
Product Details & Documents
LDPE, natural.
- Good chemical resistance
- Good thermal resistance
- Leakproof
Documents
Specifications
- Thread:32 mm
- Seq:03
- For bottles:50/100 ml