VWR® Basic Mob Caps, Detectable
Red, 530 mm
Catalog # 113-8898
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Double elastic
- Metal detectable, therefore, ideal for use in the food processing industry
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
Documents
Food Contact Certificate
Additional Product Details
TDS Mob Caps, Detectable
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Ø:530 mm
- Colour:Red
- Cat. No.:113-8898
- Garment application:General Purpose|Food
- Weight:10 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



