Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Basic Mob Caps, Detectable

VWR® Basic Mob Caps, Detectable

Blue, 530 mm
Catalog # 113-8896
Supplier:   VWR Collection
VWR® Basic Mob Caps, Detectable
Catalog # 113-8896
Supplier:  VWR Collection
VWR® Basic Mob Caps, Detectable
VWR® Basic Mob Caps, Detectable

Blue, 530 mm

Price
658,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents

These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • Free from natural rubber latex
  • Double elastic
  • Metal detectable, therefore, ideal for use in the food processing industry

Weight: 10 g/m²

Emballering: 100 pieces per bag, 10 bags per case.

Specifikationer
  • Ø:
    530 mm
  • Colour:
    Blue
  • Cat. No.:
    113-8896
  • Garment application:
    General Purpose|Food
  • Weight:
    10 g/m²
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.