[EN]IODOACETAMIDE (D4, 98%) 0,05g 1 * 0,05 g
Catalog # CAMBDLM-7249-0.05
Supplier: CAMBRIDGE ISOTOPE LABS
[EN]IODOACETAMIDE (D4, 98%) 0,05g 1 * 0,05 g
Catalog # CAMBDLM-7249-0.05
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals