Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Phenol red

100 g
Catalog # TCIAP0100-100G
Irritant
Supplier:   TCI
Phenol red
Catalog # TCIAP0100-100G
Supplier:  TCI
CAS Number:  143-74-8
Phenol red
Phenol red

100 g

CAS Number:  143-74-8
Price
739,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Merck Index:14,07247
  • Melting Pt:> 300 °C
  • Storage Temperature:Ambient
  • EINECS:205-609-7
Specifications

Specification Test Results

  • Appearance:
    Dark red to Brown powder to crystaline
  • Absorbance:
    min. 0.530 (5mg/L, pH 8.4, 556.0 to 560.0 nm)
  • Transition interval (pH):
    (yellow)6.8 to 8.4 (red)

Specifications

  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.