Order Entry
Denmark
ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 3d, solid stem, white backed, 0+120:0,5°C, length 430mm, imm. 100mm, red special liquid, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA400010-FL
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 3d, solid stem, white backed, 0+120:0,5°C, length 430mm, imm. 100mm, red special liquid, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA400010-FL
Supplier:  Amarell
Frequently Bought Together