Order Entry
Denmark
ContactUsLinkComponent
Refill therm. for silo and pit therm., -38+50:1C, special red liquid filled, milk glass scale, cap. lens front, yellow backed, lowe r part 1000mm, partial immersion 1 * 1 items
Catalog # AMARK190082-FL
Supplier:  Amarell
Refill therm. for silo and pit therm., -38+50:1C, special red liquid filled, milk glass scale, cap. lens front, yellow backed, lowe r part 1000mm, partial immersion 1 * 1 items
Catalog # AMARK190082-FL
Supplier:  Amarell
Frequently Bought Together