Order Entry
Denmark
ContactUsLinkComponent
Fadentherm., 0+100:1°C, enclosed scale, cap. colourless, red special liquid, 300xc.9mm, bulb length 100mm, partial immersion, 16 B g lass, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL44000-FL
Supplier:  Amarell
Fadentherm., 0+100:1°C, enclosed scale, cap. colourless, red special liquid, 300xc.9mm, bulb length 100mm, partial immersion, 16 B g lass, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL44000-FL
Supplier:  Amarell
Frequently Bought Together