Thermometer similar to BS 1365, SB215C/total, 205+215:0,1°C, solid stem, white backed, gallium filled, 235mm long, total immersion, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA402584-GA
Supplier: Amarell
Frequently Bought Together