Order Entry
Denmark
ContactUsLinkComponent
Thermometer similar to BS 1365, SA360C/total, 295+360:0,5°C, solid stem, white backed, gallium filled, 215mm long, total immersion, durable pigment, suitable for government verification 1 * 1 items
  AMARA402512-GA
 :  Amarell
Thermometer similar to BS 1365, SA360C/total, 295+360:0,5°C, solid stem, white backed, gallium filled, 215mm long, total immersion, durable pigment, suitable for government verification 1 * 1 items
  AMARA402512-GA
 :  Amarell
Frequently Bought Together