Order Entry
Denmark
Orders LinkContactUsLinkComponent
3-Phenoxybenzyl bromide

3-Phenoxybenzyl bromide

Bottle, 1 g
Catalog # APOSOR12353-1G
Corrosive
Irritant
Supplier:   Apollo Scientific
3-Phenoxybenzyl bromide
Catalog # APOSOR12353-1G
Supplier:  Apollo Scientific
CAS Number:  51632-16-7
3-Phenoxybenzyl bromide

Bottle, 1 g

CAS Number:  51632-16-7
Price
758,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-(Bromomethyl)-3-phenoxybenzene, 3-(Bromomethyl)diphenyl ether
  • CAS Number:51632-16-7
  • Formula:C₁₃H₁₁BrO
  • MDL Number:MFCD01761658
  • Molecular Weight:263.13 g/mol
  • Boiling Pt:112…118 °C/0.1mmHg
  • Storage Temperature:Ambient
  • EINECS:257-327-9
  • UN:3265
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications