4-Methylmorpholine
Bottle, 500 mL
Catalog # APOSOR16938-500ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: NMM, N-Methylmorpholine
- CAS Number:109-02-4
- Formula:C₅H₁₁NO
- MDL Number:MFCD00006175
- Molecular Weight:101.15 g/mol
- Boiling Pt:115…116 °C
- Melting Pt:–66 °C
- Density:0.92 g/cm³ (25 °C)
- Flash Pt:14 °C
- Storage Temperature:Ambient
- EINECS:203-640-0
- UN:2535
- ADR:3+8,II
Specifications
Specifications
- Pack type:Bottle
- Pk:500 mL
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



