Baicalein
Bottle, 5 g
Catalog # APOSOR1525T-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
An inhibitor of 12-lipoxygenase, leukotriene biosynthesis & release of lysosomal enzymes. It also inhibits cellular calcium-ion uptake, mobilization & adjuvant-induced arthirritis.
Synonyms: 5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
- CAS Number:491-67-8
- Formula:C₁₅H₁₀O₅
- MDL Number:MFCD00017459
- Molecular Weight:270.24 g/mol
- Melting Pt:264…265 °C
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



