C2 Ceramide
Bottle, 5 mg
Catalog # APOSOR6251T-5MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
A biologically active, cell permeable, non-physiological ceramide analog. It inhibits cell proliferation, induces monocytic differentiation of HL-60 cells & inducesapoptosis.
- CAS Number:3102-57-6
- Formula:C₂₀H₃₉NO₃
- MDL Number:MFCD00153903
- Molecular Weight:341.536 g/mol
- Boiling Pt:87…89 °C
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Bottle
- Pk:5 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



