Spermidine
Bottle, 5 g
APOSBIS5620-5G
Used in the isolation of chromosomes.
: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
- :124-20-9
- :C₇H₁₉N₃
- :MFCD00008229
- :145.25 g/mol
- :24 °C
- :0.925 g/cm³ (25 °C)
- :112 °C
- :Refrigerator
- :2735
- :8,II
- Pack type:Bottle
- Pk:5 g



