Order Entry
Denmark
Orders LinkContactUsLinkComponent
Apigenin 97+%

Apigenin 97+%

Bottle, 1 g
Catalog # APOSBIK9018-1G
Irritant
Supplier:   Apollo Scientific
Apigenin 97+%
Catalog # APOSBIK9018-1G
Supplier:  Apollo Scientific
CAS Number:  520-36-5
Apigenin 97+%

Bottle, 1 g

CAS Number:  520-36-5
Price
330,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
  • CAS Number:520-36-5
  • Formula:C₁₅H₁₀O₅
  • MDL Number:MFCD00006831
  • Molecular Weight:270.24 g/mol
  • Melting Pt:300 °C
  • Storage Temperature:Refrigerator
  • EINECS:208-292-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.