Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Hydroxyurea 98%

Bottle, 5 g
Catalog # APOSBIH0153-5G
Health
Supplier:   Apollo Scientific
Hydroxyurea 98%
Catalog # APOSBIH0153-5G
Supplier:  Apollo Scientific
CAS Number:  127-07-1
Hydroxyurea 98%

Bottle, 5 g

CAS Number:  127-07-1
Price
220,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Anti-neoplastic. Inactivates ribonucleoside reductase byformation of free radical nitroxide. This blocks synthesis of deoxynucleotides which inhibits DNA synthesis.

  • Protein
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Melting Pt:135 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.