Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Hydroxyurea 98%

Bottle, 25 g
Catalog # APOSBIH0153-25G
Health
Supplier:   Apollo Scientific
Hydroxyurea 98%
Catalog # APOSBIH0153-25G
Supplier:  Apollo Scientific
CAS Number:  127-07-1
Hydroxyurea 98%

Bottle, 25 g

CAS Number:  127-07-1
Price
494,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

Anti-neoplastic. Inactivates ribonucleoside reductase byformation of free radical nitroxide. This blocks synthesis of deoxynucleotides which inhibits DNA synthesis.

  • Protein
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Melting Pt:135 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications