Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(+)-Arginine

L(+)-Arginine

Bottle, 500 g
Catalog # APOSBIA0704-500G
Irritant
Supplier:   Apollo Scientific
L(+)-Arginine
Catalog # APOSBIA0704-500G
Supplier:  Apollo Scientific
CAS Number:  74-79-3
L(+)-Arginine

Bottle, 500 g

CAS Number:  74-79-3
Price
253,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

USP, pyrogen free

Synonyms:  H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
  • CAS Number:74-79-3
  • Formula:C₆H₁₄N₄O₂
  • MDL Number:MFCD00002635
  • Molecular Weight:174.2 g/mol
  • Melting Pt:222…226 °C (dec.)
  • Storage Temperature:Refrigerator
  • EINECS:200-811-1

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications