Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(+)-Arginine

L(+)-Arginine

Bottle, 1 kg
Catalog # APOSBIA0704-1KG
Irritant
Supplier:   Apollo Scientific
L(+)-Arginine
Catalog # APOSBIA0704-1KG
Supplier:  Apollo Scientific
CAS Number:  74-79-3
L(+)-Arginine

Bottle, 1 kg

CAS Number:  74-79-3
Price
450,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents

USP, pyrogen free

Synonyms:  H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
  • CAS Number:74-79-3
  • Formula:C₆H₁₄N₄O₂
  • MDL Number:MFCD00002635
  • Molecular Weight:174.2 g/mol
  • Melting Pt:222…226 °C (dec.)
  • Storage Temperature:Refrigerator
  • EINECS:200-811-1

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.