Order Entry
Denmark
ContactUsLinkComponent
SICCO Dessicator Black, panels black, one door, 2 shelves (max 8 pieces), hygrometer, tray for dessicant. Use normal pressure. Temperature resistance -20 to +70°C. External dimensions 560 x 560 x 580 mm. Usable Interior space 495 x 500 x 540 mm. Usab 1 * 1 items
  BOHLV1939-07
 :  Bohlender
undefined
SICCO Dessicator Black, panels black, one door, 2 shelves (max 8 pieces), hygrometer, tray for dessicant. Use normal pressure. Temperature resistance -20 to +70°C. External dimensions 560 x 560 x 580 mm. Usable Interior space 495 x 500 x 540 mm. Usab 1 * 1 items
  BOHLV1939-07
 :  Bohlender
Frequently Bought Together