Cartridge FlashPure Select Silica 220g, 320 mL, 300psi / 20bar, 120 mL/min, 220mg-66g, Spherical 13-17µm 30-70Å 600-800m 2 /g 1 * 6 items
Catalog # BUCH140000119
Supplier: BUCHI
Cartridge FlashPure Select Silica 220g, 320 mL, 300psi / 20bar, 120 mL/min, 220mg-66g, Spherical 13-17µm 30-70Å 600-800m 2 /g 1 * 6 items
Catalog # BUCH140000119
Supplier: BUCHI
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection