Order Entry
Denmark
ContactUsLinkComponent
Time Resolved Filter with holder 350nm BW 60nm 1 * 1 items
Catalog # MLDV6590-0081
Supplier:  Molecular Devices
Time Resolved Filter with holder 350nm BW 60nm 1 * 1 items
Catalog # MLDV6590-0081
Supplier:  Molecular Devices
Frequently Bought Together