Order Entry
Denmark
ContactUsLinkComponent
Human cord blood mononuclear cells are isolated by diluting the cord blood with balanced saline and using density gradient centrifug ation techniques. Cord blood is collected from healthy pregnant women under IRB approved informed consent and tested 1 * 25 Mou
Catalog # CB009C-1
Human cord blood mononuclear cells are isolated by diluting the cord blood with balanced saline and using density gradient centrifug ation techniques. Cord blood is collected from healthy pregnant women under IRB approved informed consent and tested 1 * 25 Mou
Catalog # CB009C-1
Frequently Bought Together