Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human bone marrow, unprocessed
Human bone marrow, unprocessed
Human unprocessed bone marrow, Fresh, Pooled, 100 mL
Catalog # BM008F-4
Human bone marrow, unprocessed
Catalog # BM008F-4
CAS Number:  
Human bone marrow, unprocessed
Human unprocessed bone marrow, Fresh, Pooled, 100 mL
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 mL
Product Details & Documents
Bone marrow is a major source of stem cells, which are being used in cancer research, cell studies, transplantation studies, and other research.
  • Test negative for Hepatitis B and C, HIV 1/2, HTLV- I/II, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
Bone marrow collected from healthy human volunteer donors under IRB approved informed consent, drawn from the posterior iliac crest using a syringe containing anticoagulant.
Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Specifications
Specifications
  • Cell Format:
    Fresh
  • Pk:
    100 mL
  • Single Donor/Pooled:
    Pooled
  • Description:
    Human unprocessed bone marrow