Order Entry
Denmark
Orders LinkContactUsLinkComponent
Colony counters

Colony counters

DOT colony counter with 120 mm magnifying glass with light
Catalog # IULA90008700
Supplier:   IUL Instruments SA
Colony counters
Catalog # IULA90008700
Supplier:  IUL Instruments SA
Colony counters
Colony counters

DOT colony counter with 120 mm magnifying glass with light

Price
9.340,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.

  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification

220 V voltage, 50 Hz, 0,2 A.

Specifications
  • Description:
    DOT colony counter with 120 mm magnifying glass with light
  • Seq:
    02