Order Entry
Denmark
Orders LinkContactUsLinkComponent
Chloroform solution, NMR reference standard, Supelco®

Chloroform solution, NMR reference standard, Supelco®

Chloroform solution, 1% in acetone-d6 (99,9 atom % D), TMS 0.1 %, NMR reference standard, NMR tube size 5 mm x 8 in., 1
Catalog # 612979-1EA
Supplier:   Merck
Chloroform solution, NMR reference standard, Supelco®
Catalog # 612979-1EA
Supplier:  Merck
CAS Number:  
Chloroform solution, NMR reference standard, Supelco®
Chloroform solution, NMR reference standard, Supelco®

Chloroform solution, 1% in acetone-d6 (99,9 atom % D), TMS 0.1 %, NMR reference standard, NMR tube size 5 mm x 8 in., 1

CAS Number:  
Price
1.310,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications

Specifications

  • Concentration:
    1% in acetone-d6 (99,9 atom % D), TMS 0.1 %
  • Pk:
    1
  • Standards:
    NMR reference standard
  • Description:
    Chloroform solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.
  • Cat. No.:
    612979-1EA
  • Description:
    Chloroform solution

Specifications